Bsu DNA Polymerase, Large fragment (from E. coli)

Lieferant: G Bioscienes

786-1999
GENO786-1999EA 386 EUR
GENO786-1999
Bsu DNA Polymerase, Large fragment (from E. coli)
Enzyme

Bsu DNA Polymerase. Large fragment is from Bacillus subtilis and is modified to retain its 5'→ 3' DNA polymerase activity with its 5'→ 3' exonuclease domain removed. It also lacks the 3'→ 5' exonuclease activity.


  • Product source: E. coli strain expressing Bsu DNA polymerase, large fragment gene
  • Storage temperature: −20 °C
  • Heat inactivation: 65 °C for 20 min


Product includes: Bsu DNA polymerase, large fragment and 10X Bsu DNA polymerase, large fragment reaction buffer


10X Bsu DNA polymerase, large fragment:
100 mM Tris-HCl
500 mM KCl
100 mM MgCl2
10 mM DTT, pH 7,9 at 25 °C


Storage buffer:
50mM Tris-HCl
50 mM KCl
1 mM DTT
0,1 mM EDTA
50% Glycerol, pH 7,5 at 25 °C


Unit definition: One unit is defined as the amount of enzyme that incorporates 10 nmoles of dNTP into acid-insoluble form in 30 minutes at 37 °C
Quality Control assays: Bsu DNA Polymerase, Large fragment is free from detectable nuclease contamination


Applications:
Second strand cDNA synthesis
Strand displacement DNA synthesis
Random primer labeling
dA-tailing

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR