Protease inhibitor cocktail, TCM-ProteaseArrest™
Lieferant: G Bioscienes
786-239
786-238
GENO786-239EA
668
EUR
GENO786-239
GENO786-238
Protease inhibitor cocktail, TCM-ProteaseArrest™
Zellkultur-Ergänzungen und -Zusätze
TCM-ProteaseArrest™ is a mixture of protease inhibitors with a broad specificity for the inhibition of serine, cysteine, aspartic (acid) proteases and aminopeptidases.
- For tissue culture medium to prevent the degradation of secreted proteins
- Store at –20 °C
TCM-ProteaseArrest™ is supplied as a 200X concentrated solution in sterile filtered DMSO. It is highly recommended to determine the appropriate dilution factor for a specific cell line, starting at a 1:200 dilution of TCM-ProteaseArrest™ to prevent harmful effects of DMSO to cell growth.