Protease inhibitor cocktail, Yeast/Fungal ProteaseArrest™
Lieferant: G Bioscienes
786-333
GENO786-333EA 184 EUR
GENO786-333
Protease inhibitor cocktail, Yeast/Fungal ProteaseArrest™
Zellkultur-Ergänzungen und -Zusätze
A broad range, 100X concentrated, ready-to-use protease inhibitor cocktail, Yeast/ Fungal ProteaseArrest™ inhibits yeast and fungal serine, cysteine and metalloproteases.
Applications include inhibition of protease activity in protein preparations of yeast and fungal lysates, protection of proteins from proteolysis in such applications as electrophoresis, purification, storage, assays, and other applications.