DNA ladder, 500 bp, DNAmark™
Lieferant: G Bioscienes
786-462
GENO786-462EA
186
EUR
GENO786-462
DNA ladder, 500 bp, DNAmark™
Elektrophorese-Reagenzien
Molekulargewichtsmarker
DNAmark™ 500 bp ladder consists of 10 DNA fragments ranging from 500 bp to 5000 bp. The 2000 bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 2000 bp are 40 ng. The 2000 bp fragment is 100 ng.
- Pre-mixed with loading dye and ready to use