BT Lab Systems Horizontal Gel Electrophoresis Set
Lieferant: G-Biosciences
BT112
GENOBT112EA 1240 EUR
GENOBT112
BT Lab Systems Horizontal Gel Electrophoresis Set
Elektrophorese-Systeme Horizontale Gelelektrophoresesysteme
BT Lab systems horizontal gel electrophoresis set's abundant buffer not only guarantees cooling effects, but also keeps the pH value stable during operation.
  • Contrasting sample background color on the tray for convenient sample loading
  • Auto-switch-off when the lid is removed
Gel sizes: 120×120 mm; 120×60 mm; 60×120 mm; 60×60 mm
Lieferumfang: Combs supplied: 1/2/3 wells×1,5 mm, 6/13 wells×1 mm; 8/18 wells×1 mm; 11/25 wells×1 mm.