BTSNM support materials, Chelex beads, 0,4 g with buffer
Lieferant: G Bioscienes
BTNM-0024
GENOBTNM-0024EA 102 EUR
GENOBTNM-0024
BTSNM support materials, Chelex beads, 0,4 g with buffer
Protein-Aufbereitungs Proteinreinigungs-Reagenzien
Chelex beads are made of a styrene divinylbenzene copolymer containing paired iminodiacetate ions, which act as chelating groups in binding polyvalent metal ions.