Anti-GST tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Genscript
A00130-40
GENSA00130-40EA 173 EUR
GENSA00130-40
Anti-GST tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Tag
Antigen: GST tag (Glutathione-S-Transferase tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR