Anti-HA tag Mouse Monoclonal Antibody
Lieferant: Genscript
A01244-100
GENSA01244-100EA 492 EUR
GENSA01244-100
Anti-HA tag Mouse Monoclonal Antibody
Antikörper
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen