Anti-beta-Amyloid Rabbit polyclonal antibody
Lieferant: Genscript
A01567-40
GENSA01567-40EA 192 EUR
GENSA01567-40
Anti-beta-Amyloid Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: ?-amyloid
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen