Anti-TUBB Mouse Monoclonal Antibody [clone: 2G7D4]

Lieferant: Genscript
A01717-40
GENSA01717-40EA 192 EUR
GENSA01717-40
Anti-TUBB Mouse Monoclonal Antibody [clone: 2G7D4]
Antikörper
Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: monoclonal
Clone: 2G7D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR