Anti-SARS-CoV-2 Nucleocapsid Protein Human Monoclonal Antibody [clone: HC2003]

Lieferant: Genscript
A02039-100
GENSA02039-100EA 625 EUR
GENSA02039-100
Anti-SARS-CoV-2 Nucleocapsid Protein Human Monoclonal Antibody [clone: HC2003]
Antikörper
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). Nucleocapsid Protein is a most abundant structure protein of the coronavirus which is associated with nucleic acid.
SARS-CoV-2 Nucleocapsid Antibody (HC2003), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.

Type: Primary
Antigen: Protein
Clonality: Monoclonal
Clone: HC2003 (monoclonal antibody)
Conjugation: Unconjugated
Epitope: Confidentiality
Host: Human
Isotype: Recombinant human IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR