AmMag™ Quatro Automation Module
Lieferant: Genscript
The AmMag Quatro is an automated solution for purification of large-scale, high quality, transfection-grade plasmids from Escherichia coli cultures.
- Ambient temperature: 18 ~ 32 ℃
- Relative humidity: ≤85%
- Altitude: ≤2000 m
The solution consists of a system controller that provides simultaneous operation of up to four independent purification modules, along with pre-filled consumables and verified protocols. The consumable kit is an all-inclusive solution that contains the samples tube, reaction tube, reagent cartridge, tip box that contains the elution tube and the waste container.
Note: Avoid exposure to sources of electromagnetic interference
Sample type: Escherichia coli - E. coli
Sample throughput: Six samples per purification module, 24 samples with a four purification module setup
Plasmid sample size: Bacteria wet pellet mass ≤1,5 g
Estimated run time: ~2 hours - depending on purification program
Sample/reaction tube: AmMag Quatro 50 ml tubes
Purification method: Magnetic beads method based on alkalinelysis
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...