LAL chromogenic endotoxin assay kit, ToxinSensor™
Lieferant: Genscript
ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.
- Highly sensitive – detect endotoxin concentration in the range of 0,005 to 1 EU/ml
- Fast – incubation period can be shortened to 14 minutes
- Reliable – colour stabiliser ensures accurate results
This method utilises a modified Limulus Amebocyte Lysate (LAL) and a synthetic colour producing substrate to detect endotoxin chromogenically.
Storage: The kit should be stored dry at room temperature for up to one month. For longer storage, the kit can be kept at 2 - 8 °C for up to one year. Do not freeze the kit or any of its components.
Lieferumfang: Kit includes endotoxin-free tips and tubes, LAL reagent water and incubation rack.
Bitte beachten Coloured samples (e.g. cell bacterial culture medium, serum or blood, etc.) can not be assayed using this kit.