Shaking water baths
Lieferant: GERBER FUNKE
3550
GERB3550EA 0 EUR
GERB3550
Shaking water baths
Bäder Schüttelwasserbäder
Shaking water bath, stainless steel. PID controller with PT-100 temperature sensor.
Lieferumfang: Supplied with temperature stand and 18 tubes.