Sand, Crystalline 0,1-0,5 mm, washed, calcinated

Lieferant: GERBER FUNKE

Synonyme: Silicium(IV)oxid, Siliciumdioxid

5460 5461 5462 5464
GERB5460EA 10.75 EUR
GERB5460 GERB5461 GERB5462 GERB5464
Sand, Crystalline 0,1-0,5 mm, washed, calcinated
Sand

Kristallin, kalziniert.

Formel: SiO₂
Molecular Weight: 60.08 g/mol
Schmelzpunkt: 1700
Dichte: 2.6
MDL: MFCD02100519
CAS-Nummer: 7631-86-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR