Indicator papers, moisture
Lieferant: GERBER FUNKE
5600
GERB5600EA 36.95 EUR
GERB5600
Indicator papers, moisture
Testpapiere Indikatorpapiere
Indicator paper for the determination of the moisture distribution in butter.