C-FLEX ULTRA PUMP TUBING, 0.500 ID, 0.750 OD, 0.125 WALL, LENGTH 50 FEET, 3 COILS 1 * 3 ST
Lieferant: Saint Gobain Life Sciences
C-FLEX ULTRA PUMP TUBING, 0.500 ID, 0.750 OD, 0.125 WALL, LENGTH 50 FEET, 3 COILS 1 * 3 ST