Advanced TOUCH, InduProof®
Lieferant: GETT GERATETECHNIK
KG17201 KG17202
GETTKG17201EA 318 EUR
GETTKG17201 GETTKG17202
Advanced TOUCH, InduProof®
Tastaturen
Rugged, water- and dust-proof silicone keyboard with integrated touchpad.
  • Rugged, durable keyboard with 104 keys and integrated mouse button
  • Comfortable and ergonomic operation also with protective gloves made of silicone or latex
  • IP 68 protection, can be submerged into liquids
  • Easy cleaning and disinfection - even under tap water or by immersion
  • Increased chemical & mechanical resistance due to special surface treatment
  • VESA-M5-boreholes (distance 75×75 mm)
  • 4 years warranty on technical function
  • Made in Germany
The InduProof® Advanced keyboardMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable