CleanType® Xtra Touch Protect Wireless Keyboards with Touchpad, Compact Layout IP68
Lieferant: GETT GERATETECHNIK
KG30219 KG30221 KG30222 KG30223
GETTKG30219EA 161 EUR
GETTKG30219 GETTKG30221 GETTKG30222 GETTKG30223
CleanType® Xtra Touch Protect Wireless Keyboards with Touchpad, Compact Layout IP68
Tastaturen
Wireless silicone keyboards with touchpad - perfect for use in medicine and industry.
  • Wireless 2,4 GHz: To be flexible in demanding surroundings
  • Precise and flat key profile due to scissor key mechanics
  • Cursor control by integrated capacitive touchpad
  • IP68 rated: The keyboard is completely water- and dustproof which allows easy, fast and deep cleaning and disinfection
  • Keylock function: To avoid missuse during the cleaning process you can disable the buttons of the keyboard by hitting the keylock button
  • Fold-out feet to tilt the keyboard for ergonomic work
  • Two year warranty
The CleanType® touch protect keyboards are high-quality compact silicone keyboards that are easy to clean thanks to their water, disinfection and dust resistance, making the keyboards ideal for use in hygiene-sensitive areas such as hospitals, clinics, medical practices, laboratories or pharmaceutical companies.
Wireless version is already available various country layouts. USB wired version currently available in DE layout. For other country layouts required please contact your Avantor sales representative.
Standards: CE, FCC; EN60601-1-2- Ed 4, EN301489-1-17+EN300328.
Zubehörinformationen: Including USB-A Dongle 2,4 GHz.
Bestellinformation: Requires 2×AAA batteries which are not included and must be ordered separately.