Hämatoxylin, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ500
GHS1128-4L
GHS1128-4LEA
283
EUR
GHS1128-4L
Hämatoxylin, Sigma-Aldrich®
Hämatoxylin-Färbelösung
Gill No. 1 formulation is used as a progressive cytology stain. Used with hematoxylin and eosin staining.
- 2 g/L certified hematoxylin
Formel:
C₁₆H₁₄O₆ Lagertemperatur: Room Temperature |
MDL:
MFCD00078111 CAS-Nummer: 517-28-2 |
|
Spezifikation Testergebnisse
Appearance (Turbidity) | Clear |
Appearance(Colour) Dark Reddish-Purple |
Conforms |
Solution | Conforms |
Conforms | Essentially free of precipitate |
24 months | Using Sigma-Aldrich Procedure No. GHS, Nuclei and compontents stain blue to black. Chromatin is clearly visible. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...