Acid hematoxylin solution

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ500

GHS280-2.5L
GHS280-2.5LEA 211 EUR
GHS280-2.5L
Acid hematoxylin solution
Acid hematoxylin solution

Gill formulation No. 2 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time. 4 g/L certified hematoxylin.

Bestellen

Spezifikation Testergebnisse

Appearance (Turbidity) Clear
Appearance(Colour)
Reddish-Purple
Conforms
Appearance (Form)
Essentially free of precipitate
Solution
Performance Conforms
Using Sigma-Aldrich Procedure No. GHS, Nuclei and compontents stain blue to black, Chromatin clearly visible
Expiration Date Period 24 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR