Aspiration station Y-shaped tubing connector
Lieferant: GILSON
F1077418
GILSF1077418EA 11.5 EUR
GILSF1077418
Aspiration station Y-shaped tubing connector
Fittings und Verbinder
The Gilson safe aspiration station Y-shaped tubing connector is for use with the Gilson safe aspiration station.
  • Connect two handles to Gilson Safe Sapiration Station station
The connector enables you to connect two handles to the station, allowing two people to use the station simultaneously.