Cooling racks
Lieferant: GLW
G01 G01S G02 G02S
GLWAG01EA 16.9 EUR
GLWAG01 GLWAG01S GLWAG02 GLWAG02S
Cooling racks
Gestelle Kühlgestelle
These cooling racks are made of polypropylene for test tubes that require continued cooling.
  • Sturdy construction
  • Autoclavable up to +121 °C
  • Cover must be ordered seperately
Remove the insert, add chrushed ice, reinsert and start working. Use in freezer down to −90° C
Bestellen
TECHNISCHE DATEN
Environmentally Preferable