Kryoboxen für Objektträger oder Mikroplatten
Lieferant: GLW
K30
GLWAK30EA 10.4 EUR
GLWAK30
Kryoboxen für Objektträger oder Mikroplatten
Boxen Kryoboxen
This slide box with 30 compartments and a fixed blade can provide a secure hold for slides or microplates.
  • Standard slides or microtitre plates (MTP)
  • Can be used into freezers down to –90 °C
  • Lid and safety seals can prevent the unauthorised access
It is suitable for storing, transporting, freezing as well as autoclaving the samples.
Lieferumfang: Supplied with snap lock lid and safety seals.