Inoculating loops, Corning® Gosselin™
Lieferant: Corning
OS10-02 OS1-01 OS1B-01 OS10B-02
612-3691EA 359 EUR
612-3691 612-3692 GOSSOS1B-01 GOSSOS10B-02
Inoculating loops, Corning® Gosselin™
Schaber Impfösen und Impfnadeln
These polystyrene inoculating loops are highly ergonomic and are color coded for easy identification.
  • Spherical or needle extremity
  • Assured sterile 10-3
Traceability information is provided on each bag.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable Umweltschonende Herstellung
Umweltzertifizierung RECs, ISO 14001 - Borre