IPTG (Isopropyl-β-D-thiogalactopyranosid)

Lieferant: GERBU BIOCHEMICALS

Synonyme: IPTG

1010.0005
GRBU1010.0005EA 0 EUR
GRBU1010.0005
IPTG (Isopropyl-β-D-thiogalactopyranosid)
IPTG (Isopropyl-β-D-thiogalactopyranosid)
Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
Schmelzpunkt: 109.5…110.5 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR