Test tubes with screw cap, sterile
Lieferant: Greiner Bio-One
164162_1000
GREI164162_1000EA 436 EUR
GREI164162_1000
Test tubes with screw cap, sterile
Röhrchen Reagenzgläser- und Probenröhrchen
Polystyrene (PS), clear.
  • Conical bottom, skirted
  • Good thermal resistance
  • Good chemical resistance
Universal tubes for a wide range of applications.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable Umweltschonende Herstellung
Umweltzertifizierung ISO 14001 (241002); ISO 50001 (241002)