Microplates, 96-well, no bottom
Lieferant: Greiner Bio-One
655000-06_40 655000_40
GREI655000-06_40EA 310 EUR
GREI655000-06_40 GREI655000_40
Microplates, 96-well, no bottom
Mikrotiterplatten HTS-Mikrotiterplatten
These 96-well plates are made from virgin polystyrene resin and moulded with no bottom.
  • Chimney-well design to help prevent cross-contamination