Deep well plates, 24-well no-bottom plates
Lieferant: Greiner Bio-One
662000-06_28
GREI662000-06_28EA 186 EUR
GREI662000-06_28
Deep well plates, 24-well no-bottom plates
Mikrotiterplatten Deep-Well-Mikroplatten
Plates are made from virgin polystyrene resin and molded without the bottom of the plate. They feature chimney-well design to help prevent cross-contamination.