Kryoboxen, CryoFreeze™
Lieferant: Greiner Bio-One
802519_20
GREI802519_20EA 468 EUR
GREI802519_20
Kryoboxen, CryoFreeze™
Boxen Kryoboxen
Polycarbonate, coloured, with clear lid.
  • Stackable
  • Fixed layout
  • Ultra low temperature storage (−196 °C)