Media Bottles, tetragonal, with screw cap
Lieferant: Greiner Bio-One
952700_24
GREI952700_24EA 120 EUR
GREI952700_24
Media Bottles, tetragonal, with screw cap
Flaschen Zellkulturflaschen
Polyethylene terephtalate (PET), ultra-clear.
  • Thermal stability from –60 to +60°C
  • With graduation
  • Sterile
  • Free of DNase/RNase
  • Non-cytotoxic and non-pyrogenic
Media bottle with screw cap for liquid storage.