Creatinin

Lieferant: GREINER BIOCHEMICAL

Synonyme: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

11711704
GRNB11711704EA 68 EUR
GRNB11711704
Creatinin
Creatinin
Formel: C₄H₇N₃O
Molecular Weight: 113.12 g/mol
MDL: MFCD00059730
CAS-Nummer: 60-27-5
EINECS: 200-466-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR