Handheld Meter for Differential Pressure and Temperature, PRO 915
Lieferant: Greisinger
486133
GRSG486133EA 0 EUR
GRSG486133
Handheld Meter for Differential Pressure and Temperature, PRO 915
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
PRO 915 is a differential pressure and temperature high class professional handheld data logger with a rich set of features, high grade robustness and operating comfort for safe and reliable use.
  • 2× universal barbed pressure ports (+/-) for measuring differential pressure
  • Pressure zero and slope user corrections
  • Pressure peak detection
  • Fast, accurate and data logger with files read out via USB - no extra software necessary
  • Easy to use and read due to backlit dot matrix/clear text display
  • Various measurement views available, including life chart
  • Acoustic alarm with high/low thresholds and optional hysteresis
Statistical functions minimum, average, maximum. It has a built-in foldable stand and magnet for flexible operation.
Shock and impact proof. IP 67 waterproof (except pressure ports). And optionally available DAkkS/ACCREDIA certificates. The device has long battery life and external power via USB.
Other features:
Input for K-type thermocouple probes
Calculation of air speed and flow rate in combination with a Pitot tube
List of favorite functions for quick access to the most used operations
Built-in foldable stand and magnet for flexible operation
Shock and impact proof
Automatic with manual or scheduled start/stop logging type and the logging intervals are 1, 5, 10, 15, 30 s/1, 2, 5, 10, 15, 20, 30 min/one hour.