Handheld Meter for Differential Pressure, PRO 211
Lieferant: Greisinger
486124 486242 486243
GRSG486124EA 0 EUR
GRSG486124 GRSG486242 GRSG486243
Handheld Meter for Differential Pressure, PRO 211
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
PRO 211 is a differential pressure high class professional handheld meter with a rich set of features, high grade robustness and operating comfort for safe and reliable use.
  • 2× universal barbed pressure ports (±) for measuring differential pressure
  • Different pressure ranges available for the best measurement accuracy in any applications
  • Pressure zero and slope user corrections
  • Pressure peak detection
  • Fast and accurate
  • Easy to use and read due to backlit dot matrix/clear text display
  • Various measurement views available, including life chart
  • Acoustic alarm with high/low thresholds and optional hysteresis
Statistical functions minimum, average, maximum. It has a built-in foldable stand and magnet for flexible operation.
Shock and impact proof. IP 67 waterproof (except pressure ports). And optionally available DAkkS/ACCREDIA certificates. The device has long battery life and external power via USB.