Digital thermo-hygrometer, GFTH 200
Lieferant: Greisinger
600249
GRSG600249EA 0 EUR
GRSG600249
Digital thermo-hygrometer, GFTH 200
Hygrometer Thermohygrometer, digital
The GFTH 200 is a digital hygrometer thermometer with low power consumption, and integrated min-/max- value memory.
  • Relative humidity, temperature and dew point in just one instrument
  • High accuracy by means of digital works calibration
  • Offset and slope correction for easy adjustment
  • Extremely low power consumption
Lieferumfang: Main instrument is supplied with battery and user manual.