Ultra-accurate digital thermometer, GMH 3710
Lieferant: Greisinger
600332
GRSG600332EA 0 EUR
GRSG600332
Ultra-accurate digital thermometer, GMH 3710
Thermometer Digitalthermometer
This reference meter is dedicated to any calibration requirements in liquids, soft media and air/gases.
  • Offset and slope input
  • Minimum/maximum value memory, 'HOLD' function
  • Freely adjustable analog output 0 to 1 V or serial interface
PT100 probe connection via four-pin miniature DIN plug.
Standards: Probe connection in acc. to DIN EN 60751.
Lieferumfang: Supplied with a nine-volt battery.