Ultra-accurate digital thermometer, GMH 3750
Lieferant: Greisinger
600335
GRSG600335EA 0 EUR
GRSG600335
Ultra-accurate digital thermometer, GMH 3750
Thermometer Digitalthermometer
This reference meter is dedicated to any calibration requirement in liquids, soft media and air/gases.
  • Offset and slope input
  • Real time clock with day, month and year
  • Optical and acoustic minimum/maximum alarm
  • Freely adjustable analogue output 0 to 1 V or serial interface
PT100 probe connection via four-pin miniature DIN plug.
Standards: Probe connection in acc. to DIN EN 60751.
Lieferumfang: Supplied with a nine volt battery.