pH/mV/°C meter, handheld, GMH 3511
Lieferant: Greisinger
604953 605021
GRSG604953EA 0 EUR
GRSG604953 GRSG605021
pH/mV/°C meter, handheld, GMH 3511
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
For comfortable measurements of pH value and temperature.
  • ORP mode allows for automatic conversion to hydrogen system electrodes
  • Temperature compensation
  • Automatic buffer detection
  • Rating function of electrode‘s quality
  • Analog output
Easier operation ensured by a menu reduced to 5 points. Minimal measuring effort because of maintenance-free gel-electrode and automatic temperature compensation.
Lieferumfang: Model GMH 3511: Meter only; electrodes and accessories sold separately. Supplied with device, battery, calibration protocol, and manual.
Model GMH 3511-Set: Supplied with GMH 3511, pH electrode GE 114, temperature probe GF 1T-T3-B-BS, buffer capsules 5×GPH 4, 5×GPH 7, 2 plastic wide mouth bottles GPF 100.