Dissolved Oxygen Meter, Handheld, G1610

Lieferant: Greisinger
610003
GRSG610003EA 0 EUR
GRSG610003
Dissolved Oxygen Meter, Handheld, G1610
Elektrochemische Messgeräte Sauerstoff-Messgeräte
Pure measurement with a focus on precision, speed and reliability packaged in a compact housing distinguish an impressive price/performance ratio.

  • Modern and functional housing
  • Backlighting
  • Waterproof (IP 65 / IP 67)
  • Durable, long battery life
  • Stability recognition, automatic adjustment to environmental air
  • Including galvanic oxygen sensor
  • Easy calibration to air at the push of a button

The new handheld measuring devices also impress with their ergonomic design, dust and water-protected design and the illuminated display. The Oxymeter with maintenance-friendly galvanised sensor is an entry-level device suitable for everyday use. Concentrations in mg/L (ppm) and saturation in percentage can be read directly without using tables. Calibration with environmental air takes place at the push of a button. Use of a GSKA protective cap is recommended for field use in bodies of ater in order to protect the membrane. 3-line display with battery status indicator, background light, protected by an unbreakable pane, overhead display at the push of a button.

Application includes freshwater and salt water aquariums, aquaculture/fish, breeding, monitoring of wells and bodies of water.

Lieferumfang: Includes device, sensor, GWOK 02 spare membrane cap and KOH 100 spare electrolyte, 2× battery, manual.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR