Residual oxygen measuring systems, ResOx 5695-H / ResOx 5695-L
Lieferant: Greisinger
610040 610041
GRSG610040EA 0 EUR
GRSG610040 GRSG610041
Residual oxygen measuring systems, ResOx 5695-H / ResOx 5695-L
Elektrochemische Messgeräte Sauerstoff-Messgeräte
These ready-to-use oxygen measuring systems with gas pump are ideal for the quality control of MAP food packaging and comparable applications.
  • With data logger and interface
  • Pressure-compensated measurement - particularly important for rigid packages
  • Membrane filter prevents unintended suction of particles / liquid
  • Independent battery-operated gas pump
  • Waterproof device and sensor (IP65, IP67)
ResOx 5695-H: With sensor GOEL 370, for gases with elevated CO2 percentage
ResOx 5695-L: With sensor GOEL 381, for generic protection gases and precise measurements also at low values
Delivery rate: Approx. 80 ml/min (with GDZ 29 Filter)
Lieferumfang: The measuring systems are supplied with sensor (GOEL 370 or GOEL 381), gas pump GS 150, battery, connection lines, hoses/T-piece, 2 filters, 2 puncture needles Ø 0,9 mm, 1× 45 pcs. adhesive seal and carrying case GKK 1420.