Giemsa's Azur-Eosin-Methylenblau, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Giemsa's-Färbemittel, Giemsa's Farbstoff, Azur-Eosin-Methylenblau nach Giemsa, Giemsa's Färbung, Giemsas Azur-Eosin-Methylenblau, Giemsa-Färbemittel, Azur-Eosin-Methylenblau Giemsa

MQ500

GS128-4L
GS128-4LEA 296 EUR
GS128-4L
Giemsa's Azur-Eosin-Methylenblau, Sigma-Aldrich®
Giemsa's Azur-Eosin-Methylenblau
Flammpunkt: 11 °C (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00081642
CAS-Nummer: 51811-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caba8232d83e17a3362d1ca2e1236171318c9bda&vid=ec3523e4f2f4665810ce1a7ceca675dbb79ee690&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Turbidity) Clear
Essentially free of precipitate
Appearance (Colour) Faint Blue to Dark Blue
Appearance (Form) Solution
pH
at 25 degrees C
6.75 - 6.95
Performance Conforms
Cells stain within the performance characteristics when using Sigma-Aldrich Procedure No. GS-10
Expiration Date Period 24 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR