Coronavirus and SARS-CoV-2 ELISA kits, GSD NovaLisa®
Lieferant: GOLD STANDARD DIAGNOSTICS
CVGQ0970
GSDCCVGQ0970EA 681 EUR
GSDCCVGQ0970
Coronavirus and SARS-CoV-2 ELISA kits, GSD NovaLisa®
Klinisches Diagnosezubehör Klinische Reagenzien und Kits
Immunoassay for the quantitative and qualitative detection of specific IgG-class antibodies to the spike protein of SARS-CoV-2.
  • Quantitative determination of antibodies to the SARS-CoV-2 spike (S) protein
  • Trimeric SARS-CoV-2 Spike antigen
  • Intended as an aid to evaluate patients immune status and determine vaccine efficacy
  • Calibrators correlate to First WHO International Standards for SARS-CoV-2
  • Diagnostic sensitivity and specificity of 99.94% and 99.49% respectively
  • Validated for serum, plasma, and finger prick (serum from capillary blood) samples
It is intended to aid in the diagnosis of patients suspected of having COVID-19 disease or asymptomatic presentation of SARS-CoV-2, to monitor antibody levels during and after a COVID-19 infection, and to determine antibody levels before and after a COVID-19 vaccination.