Coronavirus and SARS-CoV-2 direct RT real-time PCR kits, GSD NovaPrime®
Lieferant: GOLD STANDARD DIAGNOSTICS
PCOV6043
PCOV6044
GSDCPCOV6043EA
806
EUR
GSDCPCOV6043
GSDCPCOV6044
Coronavirus and SARS-CoV-2 direct RT real-time PCR kits, GSD NovaPrime®
Klinisches Diagnosezubehör
Klinische Reagenzien und Kits
This Real-Time RT-PCR assay is intended for the qualitative detection of SARS-CoV-2 from human nasopharyngeal swabs, eluted directly into deionised water without an additional RNA extraction process.
- Simplify the process for COVID-19 testing
- Meet the increasing global demand for molecular solutions
- Maximize accessibility of RT-PCR testing to a larger number of laboratories
- Increase test processing
- Reduce the cost, labour, and time required to perform RT-PCR
This new workflow shortens the complete turnaround time of the test, providing results in less than 2 hours from sample to result without compromising the high accuracy of PCR testing.
Standards: CE-IVD marked.