Real Time RT-PCR Kit
Lieferant: GOLD STANDARD DIAGNOSTICS
PCOV6111
GSDCPCOV6111EA
700
EUR
GSDCPCOV6111
Real Time RT-PCR Kit
Tests
qPCR-Assays
High quality RT-PCR kits for detection of various infectious diseases.
- Fast and reliable results in approximately 1 hour
- Validated for a variety of specimen types
- Excellent performance with high sensitivity and specificity
- Extraction/inhibition and positive control included
- Suitable for most PCR platforms