Multi-Use Cytotoxic Drug Spills Kit, Guest Medical
Lieferant: GUEST
H9612
GUESH9612EA 176 EUR
GUESH9612
Multi-Use Cytotoxic Drug Spills Kit, Guest Medical
Absorptionsmaterial
The kit contains everything required for both liquid and powder spillages in one case that can be carried to the site where it is needed.
  • 2 shakers of CT-Zorb Granules
  • 3 Hazard Scoops and Scrapers
  • 3 disposable tweezers
  • 3 absorbent pads
  • 3 sterile saline pods
  • 6 detergent wipes
  • 3 cytotoxic waste disposal bags
  • 3 bag ties
  • 3 pairs long cuff nitrile gloves
  • 3 protective disposable gowns
  • 3 pairs overshoes
  • 3 FFP3 face masks
  • Reusable goggles (BS EN 166 conformant)
  • Warning sign
  • Full instructions for use
The kit contains inert CT-Zorb Granules which will quickly and efficiently absorb all the moisture of a liquid spillage, thus containing the spill and preventing it spreading further. The granule-spill mixture can then be easily collected using the Guest Medical Scoops and Scrapers for disposal according to local procedure or regulations. Absorbent pads provide a simple method for collecting powder spillages when used with the sterile saline pods provided in the kit.