EZ Yeast™ Plasmid Prep kit
Lieferant: G Bioscienes
GZ-02
GZ-02EA 468 EUR
GZ-02
EZ Yeast™ Plasmid Prep kit
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The EZ Yeast™ Plasmid Prep Kit is designed for the rapid isolation of plasmids from yeast patches or yeast grown in small liquid cultures. The kit is adapted from the alkaline lysis of E. coli, by providing modified alkaline lysis buffers and LongLife™ Zymolyase®, a highly stable enzyme.
  • Minimise time to analyse yeast plasmids; no need to shuttle plasmids to E.coli
  • Yield of 0,1 to 0,3 ng per 1,5 ml culture
  • No requirement for glass beads, phenol or excessive vortexing
  • Suitable for colonies, patches and liquid culture
  • Generates plasmids suitable for multiple downstream applications