Ethyl-4,4-difluorocyclohexanecarboxylate 97%

Lieferant: Thermo Scientific

Synonyme: Ethyl 4,4-difluorocyclohexanecarboxylate

H25754.06
H25754.06EA 250 EUR
H25754.06
Ethyl-4,4-difluorocyclohexanecarboxylate 97%
Ethyl-4,4-difluorocyclohexanecarboxylate

Purity: 97% CAS No.: 178312-47-5 Molecular Formula: C9H14F2O2 Formula Weight: 192.21 Boiling Point: 84-86°/8mm MDL No.: MFCD03788494

Formel: C₉H₁₄F₂O₂
Molecular Weight: 192,21 g/mol
Siedepunkt: 84…86 °C (8 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03788494
CAS-Nummer: 178312-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR