4-Fluoropiperidine hydrochloride 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H25755.03
H25755.MD
H25755.03EA
316
EUR
H25755.03
H25755.MD
4-Fluoropiperidine hydrochloride 95%
4-Fluoropiperidine hydrochloride
Purity: 95%
CAS No.: 57395-89-8
Molecular Formula: C5H10FN.HCl
Formula Weight: 139.60
Melting Point: 161-163°
MDL No.: MFCD03452786
Formel:
C₅H₁₁ClFN Molecular Weight: 139.6 g/mol Schmelzpunkt: 161…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03452786 CAS-Nummer: 57395-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...