3-Brom-5-fluorpyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

H25766.03 H25766.06
H25766.03EA 34.1 EUR
H25766.03 H25766.06
3-Brom-5-fluorpyridin 97%
3-Bromo-5-fluoropyridine

Purity: 99% CAS No.: 407-20-5 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Flash Point: 64°(148°F) Odour: Characteristic UN No.: 2811 MDL No.: MFCD04112555

Formel: C₅H₃BrFN
Schmelzpunkt: 24…28 °C
Flammpunkt: 64 °C (148 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD04112555
CAS-Nummer: 407-20-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR