2-Hydroxy-5-nitro-6-picolin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H25770.06
H25770.06EA 146 EUR
H25770.06
2-Hydroxy-5-nitro-6-picolin ≥98%
2-Hydroxy-5-nitro-6-picolin

Purity: 98% CAS No.: 28489-45-4 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: 233-236° MDL No.: MFCD00092010

Formel: C₆H₆N₂O₃
Schmelzpunkt: 233…236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092010
CAS-Nummer: 28489-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR